missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Methoxyquinoline, 96%

Manufacturer:  Alfa Aesar™ H54430MD

 View more versions of this product

Catalog No. AAH54430MD



Description

Description

Applications
8-Methoxyquinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

8-Methoxyquinoline
250mg
C10H9NO
4y56u1unuj
ZLKGGEBOALGXJZ-UHFFFAOYSA-N
8-methoxyquinoline
70310
96%
38°C to 41°C
938-33-0
MFCD00957068
Slightly soluble in water.
COC1=CC=CC2=C1N=CC=C2
159.188
159.19

Chemical Identifiers

938-33-0
159.188
ZLKGGEBOALGXJZ-UHFFFAOYSA-N
70310
COC1=CC=CC2=C1N=CC=C2
C10H9NO
MFCD00957068
4y56u1unuj
8-methoxyquinoline
Safety and Handling

Safety and Handling

EINECSNumber : 213-341-7

TSCA : No

Recommended Storage : Ambient temperatures

Documents