missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Methoxyquinoline-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5444503

 View more versions of this product

Catalog No. AAH5444503



Specifications

Specifications

8-Methoxyquinoline-5-boronic acid pinacol ester
916162-54-4
MFCD20527214
UJPLJBDGXPKCDH-UHFFFAOYSA-N
8-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
71422855
96%
1g
C16H20BNO3
2-8-methoxy-5-quinolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC=NC3=C(C=C2)OC
285.15
285.15

Chemical Identifiers

916162-54-4
285.15
UJPLJBDGXPKCDH-UHFFFAOYSA-N
71422855
B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC=NC3=C(C=C2)OC
C16H20BNO3
MFCD20527214
2-8-methoxy-5-quinolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
8-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Documents