missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Methoxy-2-methylquinoline 98.0+%, TCI America™

Manufacturer:  TCI America M10885G

 View more versions of this product

Catalog No. M10885G



Specifications

Specifications

8-Methoxy-2-methylquinoline
124°C
3033-80-5
MFCD00034436
OQXVXPBDSJQYRR-UHFFFAOYSA-N
8-methoxy-2-methylquinoline
316986
Crystal-Powder at 20°C
White-Yellow
5g
C11H11NO
8-Methoxyquinaldine
CC1=NC2=C(C=CC=C2OC)C=C1
173.215
173.22
≥98.0% (GC,T)

Chemical Identifiers

3033-80-5
173.215
OQXVXPBDSJQYRR-UHFFFAOYSA-N
316986
CC1=NC2=C(C=CC=C2OC)C=C1
C11H11NO
MFCD00034436
8-Methoxyquinaldine
8-methoxy-2-methylquinoline
Safety and Handling

Safety and Handling

TSCA : No

Documents