Please login to your online account to display your discounted pricing

8-M-PDOT, Tocris

Melatonin agonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material 8-M-PDOT
Assay Percent Range >99%
Molecular Formula C14H19NO2
Formula Weight 233.31
Synonym 8-Methoxy-2-propionamidotetralin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

103550

 
r & d systems
RDS103550
50mg Each for $762.58

103510

 
r & d systems
RDS103510
10mg Each for $181.19
Description & Specifications

Specifications

Formula Weight 233.31
Quantity 50mg
Chemical Name or Material 8-M-PDOT
Assay Percent Range >99%
Molecular Formula C14H19NO2
CAS 134865-70-6
Synonym 8-Methoxy-2-propionamidotetralin
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature