missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
8-Hydroxyquinoline TS, (U.S.P. Test Solution), Spectrum™
H-163, 148-24-3
Chemical Identifiers
CAS
148-24-3
Molecular Formula
C9H7NO
Molecular Weight (g/mol)
145.16
InChI Key
MCJGNVYPOGVAJF-UHFFFAOYSA-N
IUPAC Name
quinolin-8-ol
SMILES
OC1=C2N=CC=CC2=CC=C1
View More Specs
Products
2
Catalog Number
Mfr. No.
Quantity
Packaging
Price
Quantity
18600144
View Documents
Spectrum Chemical Mfg Cor
H163100ML
100 mL
Poly Bottle
Each for $97.44
Add to cart
18600145
View Documents
Spectrum Chemical Mfg Cor
H163500ML
500 mL
Poly Bottle
Each for $155.42
Add to cart
Description
Description
Spectrum™ 8-Hydroxyquinoline TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Specifications
Chemical Identifiers
CAS
148-24-3
Molecular Weight (g/mol)
145.16
IUPAC Name
quinolin-8-ol
Molecular Formula
C9H7NO
InChI Key
MCJGNVYPOGVAJF-UHFFFAOYSA-N
SMILES
OC1=C2N=CC=CC2=CC=C1
Specifications
Packaging
Poly Bottle
CAS
148-24-3
Molecular Formula
C9H7NO
InChI Key
MCJGNVYPOGVAJF-UHFFFAOYSA-N
IUPAC Name
quinolin-8-ol
Grade
Chemical Solution
Quantity
100 mL
CAS Max %
1
UN Number
UN1170
SMILES
OC1=C2N=CC=CC2=CC=C1
Molecular Weight (g/mol)
145.16
SDS