missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Hydroxyquinoline sulfate monohydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0274322
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
8-Hydroxyquinoline sulfate monohydrate | |
176°C to 179°C | |
MFCD00149610 | |
3760550 | |
BNCXJZDIJIVJJO-UHFFFAOYSA-N | |
406.42 (388.40 Anhydrous) |
207386-91-2 | |
C18H18N2O7S | |
100 g | |
8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard | |
406.41 | |
99% |
Chemical Identifiers
207386-91-2 | |
406.41 | |
BNCXJZDIJIVJJO-UHFFFAOYSA-N |
C18H18N2O7S | |
MFCD00149610 | |
8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 205-137-1
RTECSNumber : VC8260000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only