missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Hydroxyquinoline Solution, LabChem™

$105.29 - $105.29

Chemical Identifiers

CAS 7732-18-5,64-19-7,148-24-3
CAS Max % 91.76,6.24,2
Linear Formula HOC6H3N:CHCH:CH
Formula Weight 145.16
Chemical Name or Material 8-Hydroxyquinoline Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC155801 Lab Chem
LC155801
500mL Each for $105.29
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,64-19-7,148-24-3
HOC6H3N:CHCH:CH
8-Hydroxyquinoline Solution
91.76,6.24,2
145.16

Specifications

145.16
Carbon monoxide; Carbon dioxide; Nitrogen oxides
64-19-7,148-24-3,7732-18-5
1.1cSt
Clear yellow
1.01g/mL
500mL
Passes Test
Room Temperature
Amber Glass
6.24,2,91.76
1.01g/mL
Soluble in water
Liquid
HOC6H3N:CHCH:CH
8-Hydroxyquinoline Solution

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning