Please login to your online account to display your discounted pricing

8-Hydroxyquinoline (Certified ACS), Fisher Chemical


Quantity 100g
Packaging Glass Bottle

Manufacturer: fisher scientific company  O261100

 View more versions of this product

Catalog No. O261-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 72°C
Formula Weight 145.16
Boiling Point 267°C
Purity Grade Certified ACS Grade
Quantity 100g
Physical Form Solid
Color White Beige Pale Yellow
Alpha Vector Hydroxyquinoline
Ignition Residue 0.05% max.
Synonym 8-Quinolinol
Insoluble in Alcohol 0.05% max.
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Possible risks of irreversible effects. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:1
MDL Number MFCD00006807
Sulfate 0.02% max.
Identification Pass Test
Packaging Glass Bottle
Additional Information Vapor Pressure: 4.7hPa at 100°C
Suitability Pass Test
Chemical Name or Material 8-Hydroxyquinoline
ChemAlert Storage Symbol Gray
Linear Formula HOC6H3N:CHCH:CH
CAS 148-24-3
Molecular Formula C9H7NO

Specifications

Chemical Name or Material 8-Hydroxyquinoline
CAS 148-24-3
Purity Grade Certified ACS Grade
Molecular Formula C9H7NO
Linear Formula HOC6H3N:CHCH:CH
Formula Weight 145.16
MDL Number MFCD00006807
Synonym 8-Quinolinol
Accessories
 

Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Possible risks of irreversible effects. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:1

WARNING!

SDS & Safety Documents