missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, ACS

A bacteriostatic and fungistatic agent, also a chelating agent for metals | CAS: 148-24-3 | C9H7NO | 145.16 g/mol

Supplier:  Thermo Scientific Chemicals 04127230

 View more versions of this product

Catalog No. AA4127230


Only null left
Explore available promotions

Description

Description

A bacteriostatic and fungistatic agent. Also a chelating agent for metals.8-Hydroxyquinoline is used as an antiseptic and disinfectant. Its alcoholic solution is used in liquid bandages. It is used to prepare tris(8-hydroxyquinolinato)aluminum, which finds application as a component of organic light-emitting diodes (OLED's). It is also used as a reagent and as a stabilizer of hydrogen peroxide in a rocket fuel oxidizer. It is used as a chelating agent and used as a carrier for radio-indium for diagnostic purposes. Its halogenated derivatives are used as anti-infective agents and oral antiamebics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

8-Hydroxyquinoline
72°C to 76°C
267°C
MFCD00006807
114512
14,4843
Almost insoluble in water and diethyl ether; Highly soluble in ethanol,acetone,chloroform,and benzene;Soluble in acetic acid,ethanol,acetone,chloroform,benzene and aqueous mineral acids; Insoluble in water and diethylether.
OC1=C2N=CC=CC2=CC=C1
145.16
CHEBI:48981
ACS
148-24-3
White to Yellow
C9H7NO
250 g
Light sensitive
8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
MCJGNVYPOGVAJF-UHFFFAOYSA-N
quinolin-8-ol
1923
145.16
Granules

Chemical Identifiers

148-24-3
145.16
MCJGNVYPOGVAJF-UHFFFAOYSA-N
1923
quinolin-8-ol
C9H7NO
MFCD00006807
8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
CHEBI:48981
OC1=C2N=CC=CC2=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.

P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c

H301-H317-H318-H360

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 205-711-1

RTECSNumber : VC4200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only