Please login to your online account to display your discounted pricing

8-Hydroxyquinoline, ACS Grade, LabChem™

$161.49 - $161.49

Chemical Identifiers

Chemical Name or Material 8-Hydroxyquinoline
CAS 148-24-3
CAS Max % 100
Purity Grade ACS Grade
Linear Formula HOC6H3N:CHCH:CH
Formula Weight 145.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC155759

 
lab chem
LC155759
Amber Glass 100g Each for $161.49
Description & Specifications

Specifications

Formula Weight 145.16
Chemical Name or Material 8-Hydroxyquinoline
EINECS Number 205-711-1
Packaging Amber Glass
Purity Grade ACS Grade
Physical Form Crystals
Solubility Soluble in water
Linear Formula HOC6H3N:CHCH:CH
Storage Information Room Temperature
CAS 148-24-3
Suitability Passes Test
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Wash exposed skin thoroughly after handling.
Store locked up in a well-ventilated place.
Keep container tightly closed.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Carbon dioxide; Carbon monoxide; Nitrogen oxides
Identification Passes Test
Boiling Point 267°C
Color White
Quantity 100g
Safety and Handling

Warning

GHS P Statement
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Wash exposed skin thoroughly after handling.
Store locked up in a well-ventilated place.
Keep container tightly closed.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.