missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Hydroxyquinoline-7-carboxylic Acid, TCI America™

$394.17 - $1,292.53

Chemical Identifiers

CAS 19829-79-9
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00671539
InChI Key JYIAZVFJRYLCBH-UHFFFAOYSA-N
PubChem CID 231363
IUPAC Name 8-hydroxyquinoline-7-carboxylic acid
SMILES C1=CC2=C(C(=C(C=C2)C(=O)O)O)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08775G View Documents TCI America
H08775G
5g Each for $394.17
 
H087725G View Documents TCI America
H087725G
25g Each for $1,292.53
 
Specifications

Chemical Identifiers

19829-79-9
189.17
JYIAZVFJRYLCBH-UHFFFAOYSA-N
8-hydroxyquinoline-7-carboxylic acid
C10H7NO3
MFCD00671539
231363
C1=CC2=C(C(=C(C=C2)C(=O)O)O)N=C1

Specifications

Undesignated
19829-79-9
MFCD00671539
C1=CC2=C(C(=C(C=C2)C(=O)O)O)N=C1
189.17
189.17
≥98.0% (GC,T)
5g
C10H7NO3
JYIAZVFJRYLCBH-UHFFFAOYSA-N
8-hydroxyquinoline-7-carboxylic acid
231363
Crystal-Powder at 20°C
8-Hydroxyquinoline-7-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No