missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Hydroxyquinoline-7-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America H087725G
Specifications
8-Hydroxyquinoline-7-carboxylic Acid | |
C10H7NO3 | |
25 g | |
C1=CC2=C(C(=C(C=C2)C(=O)O)O)N=C1 | |
189.17 | |
189.17 | |
Crystalline Powder |
19829-79-9 | |
MFCD00671539 | |
JYIAZVFJRYLCBH-UHFFFAOYSA-N | |
8-hydroxyquinoline-7-carboxylic acid | |
231363 | |
≥98.0% (GC,T) |
Chemical Identifiers
19829-79-9 | |
189.17 | |
JYIAZVFJRYLCBH-UHFFFAOYSA-N | |
8-hydroxyquinoline-7-carboxylic acid |
C10H7NO3 | |
MFCD00671539 | |
231363 | |
C1=CC2=C(C(=C(C=C2)C(=O)O)O)N=C1 |
Safety and Handling
TSCA : No