missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 121925000

 View more versions of this product

Catalog No. AC121925000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

8-Hydroxyquinoline-5-sulfonic acid monohydrate
Plastic bottle
>300.0°C
283158-18-9
C9H9NO5S
Solubility in water: freely soluble. Other solubilities: slightly soluble in most common organic solvents
C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O
243.233
243.24
98%
Authentic
Yellow
500g
98%
5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxy-5-quinolinesulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150, unii-n85kc14z44
WUXYGIQVWKDVTJ-UHFFFAOYSA-N
8-hydroxyquinoline-5-sulfonic acid;hydrate
2723649
Powder

Chemical Identifiers

283158-18-9
243.233
5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxy-5-quinolinesulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150, unii-n85kc14z44
8-hydroxyquinoline-5-sulfonic acid;hydrate
C9H9NO5S
WUXYGIQVWKDVTJ-UHFFFAOYSA-N
2723649
C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. Suspected of causing genetic defects.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Use personal protective equipment as required. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Obtain special instructions before use. Avoid breathing dust/fume/gas/mist/vapors/spray.

Warning

SDS Documents