missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 121920050

 View more versions of this product

Catalog No. AC121920050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

8-Hydroxyquinoline-5-sulfonic acid monohydrate
Glass Bottle
>300.0°C
283158-18-9
C9H7NO4S·H2O
14,4844
Solubility in water: freely soluble. Other solubilities: slightly soluble in most common organic solvents
C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O
243.24
243.24
98%
Authentic
Yellow
5g
97.5% min
22,407
5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxy-5-quinolinesulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150, unii-n85kc14z44
WUXYGIQVWKDVTJ-UHFFFAOYSA-N
8-hydroxyquinoline-5-sulfonic acid;hydrate
2723649
Powder

Chemical Identifiers

283158-18-9
243.24
5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxy-5-quinolinesulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150, unii-n85kc14z44
8-hydroxyquinoline-5-sulfonic acid;hydrate
C9H7NO4S·H2O
WUXYGIQVWKDVTJ-UHFFFAOYSA-N
2723649
C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lense

GHS Signal Word: Warning

SDS Documents