missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline-5-sulfonic acid hydrate, 98%

CAS: 207386-92-3 | C9H7NO4S | 225.22 g/mol

Supplier:  Thermo Scientific Chemicals B2002236

 View more versions of this product

Catalog No. AAB2002236


Only null left
Explore available promotions

Description

Description

It is employed as a reagent for determination of trace metals. It is employed as pre-1,2 and post-column3 reagent for fluorescence detection of metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reagent for determination of trace metals. It is employed as pre-1,2 and post-column3 reagent for fluorescence detection of metal ions.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

8-Hydroxyquinoline-5-sulfonic acid hydrate
311°C to 313°C
MFCD00149482
195247
8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, unii-n85kc14z44, 8-hydroxy-5-quinolinesulfonic acid hydrate, 5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150
LGDFHDKSYGVKDC-UHFFFAOYSA-N
8-hydroxyquinoline-5-sulfonic acid;hydrate
2723649
98%
207386-92-3
C9H7NO4S
500 g
14,4844
Soluble in water.
OC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O
225.22
225.22 (Anhydrous)

Chemical Identifiers

207386-92-3
225.22
LGDFHDKSYGVKDC-UHFFFAOYSA-N
2723649
OC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O
C9H7NO4S
MFCD00149482
8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, unii-n85kc14z44, 8-hydroxy-5-quinolinesulfonic acid hydrate, 5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150
8-hydroxyquinoline-5-sulfonic acid;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 201-570-5

RTECSNumber : VC2570000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only