Please login to your online account to display your discounted pricing

8-Hydroxyquinoline, 2% (w/v), Ricca Chemical

Chemical Identifiers

Chemical Name or Material 8-Hydroxyquinoline
CAS 7732-18-5
CAS Min % 90.0
CAS Max % 94.0
CAS 64-19-7
CAS Min % 5.7
CAS Max % 5.93
CAS 148-24-3
CAS Min % 1.81
CAS Max % 1.88
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3940-16

 
ricca chemical company
RI394016
Glass amber 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3940-4

 
ricca chemical company
RI39404
Glass amber 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear yellow
Physical Form Liquid
Quantity 500mL
CAS 7732-18-5
CAS Min % 90.0
CAS 64-19-7
Packaging Glass amber
CAS Max % 5.93
CAS 148-24-3
CAS Min % 1.81
Chemical Name or Material 8-Hydroxyquinoline
CAS Max % 1.88
Concentration or Composition Notes 2% (w/v)
CAS Max % 94.0
CAS Min % 5.7