Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Hydroxyquinoline-2-carboxylic acid, 98%

$133.96 - $471.62

Chemical Identifiers

Chemical Name or Material 8-Hydroxyquinoline-2-carboxylic acid
CAS 1571-30-8
Molecular Formula C10H7NO3
Formula Weight 189.17
MDL Number MFCD00168972
Synonym 8-Hydroxyquinaldic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2705403

  SDS
alfa aesar™
H2705403
1g Each for $133.96

AAH2705406

  SDS
alfa aesar™
H2705406
5g Each for $471.62
Description & Specifications

Specifications

Formula Weight 189.17
Chemical Name or Material 8-Hydroxyquinoline-2-carboxylic acid
Assay 98%
Molecular Formula C10H7NO3
MDL Number MFCD00168972
CAS 1571-30-8
Synonym 8-Hydroxyquinaldic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UZ9450000
Melting Point 216° to 221°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.