missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Hydroxyquinaldine, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 121875000
Specifications
8-Hydroxyquinaldine | |
Authentic | |
267°C | |
70°C to 74°C | |
98% | |
97.5 | |
98% | |
MFCD00006765 | |
2-methyl 8-quinolonol, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 2-methyl-quinolin-8-ol, 2-methyloxine, 8-hydroxy-2-methylquinoline, 8-hydroxyqinaldine, 8-hydroxyquinaldine, 8-quinolinol, 2-methyl, hydroxyquinaldine | |
NBYLBWHHTUWMER-UHFFFAOYSA-N | |
2-methylquinolin-8-ol | |
13224 | |
Crystalline Powder |
139°C | |
Glass bottle | |
Beige to Brown | |
500g | |
826-81-3 | |
100.0 | |
C10H9NO | |
21, 106 | |
Solubility in water: insoluble | |
CC1=NC2=C(C=CC=C2O)C=C1 | |
159.19 | |
159.19 | |
98% |
Chemical Identifiers
826-81-3 | |
159.19 | |
NBYLBWHHTUWMER-UHFFFAOYSA-N | |
13224 | |
CC1=NC2=C(C=CC=C2O)C=C1 |
C10H9NO | |
MFCD00006765 | |
2-methyl 8-quinolonol, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 2-methyl-quinolin-8-ol, 2-methyloxine, 8-hydroxy-2-methylquinoline, 8-hydroxyqinaldine, 8-hydroxyquinaldine, 8-quinolinol, 2-methyl, hydroxyquinaldine | |
2-methylquinolin-8-ol |
Safety and Handling
EINECSNumber : 212-562-6
RTECSNumber : VC7920000
TSCA : TSCA