missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Hydroxyquinaldine, 98%, ACROS Organics™

$23.64 - $23.64

Chemical Identifiers

CAS 826-81-3
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00006765
InChI Key NBYLBWHHTUWMER-UHFFFAOYSA-N
Synonym 2-methyl 8-quinolonol, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 2-methyl-quinolin-8-ol, 2-methyloxine, 8-hydroxy-2-methylquinoline, 8-hydroxyqinaldine, 8-hydroxyquinaldine, 8-quinolinol, 2-methyl, hydroxyquinaldine
PubChem CID 13224
IUPAC Name 2-methylquinolin-8-ol
SMILES CC1=NC2=C(C=CC=C2O)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC121870250
SDS View Documents
Fisher Scientific Acros
121870250
25g Glass bottle
Each for $23.64
 
AC121871000
SDS View Documents
Thermo Fisher Scientific
121871000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC121875000
SDS View Documents
Thermo Fisher Scientific
121875000
500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

826-81-3
159.19
NBYLBWHHTUWMER-UHFFFAOYSA-N
13224
CC1=NC2=C(C=CC=C2O)C=C1
C10H9NO
MFCD00006765
2-methyl 8-quinolonol, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 2-methyl-quinolin-8-ol, 2-methyloxine, 8-hydroxy-2-methylquinoline, 8-hydroxyqinaldine, 8-hydroxyquinaldine, 8-quinolinol, 2-methyl, hydroxyquinaldine
2-methylquinolin-8-ol

Specifications

139°C
Glass bottle
Beige to Brown
25g
97.5
98%
MFCD00006765
2-methyl 8-quinolonol, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 2-methyl-quinolin-8-ol, 2-methyloxine, 8-hydroxy-2-methylquinoline, 8-hydroxyqinaldine, 8-hydroxyquinaldine, 8-quinolinol, 2-methyl, hydroxyquinaldine
NBYLBWHHTUWMER-UHFFFAOYSA-N
2-methylquinolin-8-ol
13224
Crystalline Powder
8-Hydroxyquinaldine, 98%
Authentic
267°C
70°C to 74°C
826-81-3
100.0
C10H9NO
21, 106
Solubility in water: insoluble
CC1=NC2=C(C=CC=C2O)C=C1
159.19
159.19
98%
Safety and Handling

Safety and Handling

EINECSNumber : 212-562-6

RTECSNumber : VC7920000

TSCA : TSCA

SDS