missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Hydroxyjulolidine, TCI America™

$76.39 - $262.78

Chemical Identifiers

CAS 41175-50-2
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00006918
InChI Key FOFUWJNBAQJABO-UHFFFAOYSA-N
Synonym 8-hydroxyjulolidine, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinolin-8-ol, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinolin-8-ol, 8-hydroxy julolidine, timtec-bb sbb006615, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 1-azatricyclo 7.3.1.0^ 5,13 trideca-5 13 ,6,8-trien-6-ol
PubChem CID 170474
SMILES C1CC2=C3C(=C(C=C2)O)CCCN3C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09281G View Documents TCI America
H09281G
1g Each for $76.39
 
H09285G View Documents TCI America
H09285G
5g Each for $262.78
 
Specifications

Chemical Identifiers

41175-50-2
189.258
FOFUWJNBAQJABO-UHFFFAOYSA-N
170474
C12H15NO
MFCD00006918
8-hydroxyjulolidine, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinolin-8-ol, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinolin-8-ol, 8-hydroxy julolidine, timtec-bb sbb006615, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 1-azatricyclo 7.3.1.0^ 5,13 trideca-5 13 ,6,8-trien-6-ol
C1CC2=C3C(=C(C=C2)O)CCCN3C1

Specifications

Undesignated
1g
C12H15NO
8-hydroxyjulolidine, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinolin-8-ol, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinolin-8-ol, 8-hydroxy julolidine, timtec-bb sbb006615, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 1-azatricyclo 7.3.1.0^ 5,13 trideca-5 13 ,6,8-trien-6-ol
C1CC2=C3C(=C(C=C2)O)CCCN3C1
170474
Crystal-Powder at 20°C
8-Hydroxyjulolidine
132°C
41175-50-2
MFCD00006918
FOFUWJNBAQJABO-UHFFFAOYSA-N
189.258
189.26
≥97.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : Yes