Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Hydroxyjulolidine, 97%

$84.53 - $268.20

Chemical Identifiers

Chemical Name or Material 8-Hydroxyjulolidine
CAS 41175-50-2
Molecular Formula C12H15NO
Formula Weight 189.26
MDL Number MFCD00006918
Beilstein 1530423
Synonym 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1769203

  SDS
alfa aesar™
A1769203
1g Each for $84.53

AAA1769206

  SDS
alfa aesar™
A1769206
5g Each for $268.20
Description & Specifications

Specifications

Formula Weight 189.26
TSCA TSCA
Health Hazard 1 Warning
Chemical Name or Material 8-Hydroxyjulolidine
Assay 97%
Molecular Formula C12H15NO
MDL Number MFCD00006918
CAS 41175-50-2
Beilstein 1530423
Synonym 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 132° to 135°C
Quantity 1g

Applications
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.

Solubility
Soluble in water (partly), ethanol, chloroform, methanol, and acetone.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.