Please login to your online account to display your discounted pricing

8-Hydroxy-PIPAT oxalate, Tocris


High affinity 5-HT1A agonist

Quantity 50mg

Manufacturer: r & d systems  RDS079750

 View more versions of this product

Catalog No. 079750

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 461.3
Chemical Name or Material 8-Hydroxy-PIPAT oxalate
Assay Percent Range >98%
Molecular Formula C16H22INO·C2H2O4
CAS 1451210-48-2
Synonym (RS)-trans-8-Hydroxy-2-[N-n-propyl-N-(3'-iodo-2'-propenyl)amino]tetralin oxalate
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material 8-Hydroxy-PIPAT oxalate
Assay Percent Range >98%
Molecular Formula C16H22INO·C2H2O4
Formula Weight 461.3
Synonym (RS)-trans-8-Hydroxy-2-[N-n-propyl-N-(3'-iodo-2'-propenyl)amino]tetralin oxalate
Accessories

Desiccate at -20°C