Please login to your online account to display your discounted pricing

8-Hydroxy-PIPAT oxalate, Tocris

High affinity 5-HT1A agonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material 8-Hydroxy-PIPAT oxalate
Assay Percent Range >98%
Molecular Formula C16H22INO·C2H2O4
Formula Weight 461.3
Synonym (RS)-trans-8-Hydroxy-2-[N-n-propyl-N-(3'-iodo-2'-propenyl)amino]tetralin oxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

079750

 
r & d systems
RDS079750
50mg Each for $895.76

079710

 
r & d systems
RDS079710
10mg Each for $214.12
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 461.3
Molecular Formula C16H22INO·C2H2O4
Assay Percent Range >98%
Chemical Name or Material 8-Hydroxy-PIPAT oxalate
CAS 1451210-48-2
Synonym (RS)-trans-8-Hydroxy-2-[N-n-propyl-N-(3'-iodo-2'-propenyl)amino]tetralin oxalate
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C