Please login to your online account to display your discounted pricing

8-Hydroxy-DPAT hydrobromide, Tocris


Selective 5-HT1A agonist. Also has moderate affinity for 5-HT7

Quantity 10mg

Manufacturer: r & d systems  RDS052910

 View more versions of this product

Catalog No. 052910

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 328.29
Assay Percent Range >98%
Molecular Formula C16H25NO·HBr
Synonym (±)-8-Hydroxy-2-dipropylaminotetralin hydrobromide
Chemical Name or Material 8-Hydroxy-DPAT hydrobromide
CAS 76135-31-4
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material 8-Hydroxy-DPAT hydrobromide
Assay Percent Range >98%
Molecular Formula C16H25NO·HBr
Formula Weight 328.29
Synonym (±)-8-Hydroxy-2-dipropylaminotetralin hydrobromide
Accessories

Desiccate at +4°C