Please login to your online account to display your discounted pricing

8-Hydroxy-DPAT hydrobromide, Tocris

Selective 5-HT1A agonist. Also has moderate affinity for 5-HT7

$88.05 - $323.08

Chemical Identifiers

Chemical Name or Material 8-Hydroxy-DPAT hydrobromide
Assay Percent Range >98%
Molecular Formula C16H25NO·HBr
Formula Weight 328.29
Synonym (±)-8-Hydroxy-2-dipropylaminotetralin hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

052950

 
r & d systems
RDS052950
50mg Each for $323.08

052910

 
r & d systems
RDS052910
10mg Each for $88.05
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 328.29
Chemical Name or Material 8-Hydroxy-DPAT hydrobromide
CAS 76135-31-4
Assay Percent Range >98%
Synonym (±)-8-Hydroxy-2-dipropylaminotetralin hydrobromide
Molecular Formula C16H25NO·HBr
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C