missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Hydroxy-7-propylquinoline 98.0+%, TCI America™
$323.66 - $1,088.23
Chemical Identifiers
CAS | 58327-60-9 |
---|---|
Molecular Formula | C12H13NO |
Molecular Weight (g/mol) | 187.242 |
MDL Number | MFCD00059755 |
InChI Key | OKPSVYHYNXVYED-UHFFFAOYSA-N |
Synonym | 7-Propyloxine, 7-Propyl-8-quinolinol |
PubChem CID | 11604513 |
IUPAC Name | 7-propylquinolin-8-ol |
SMILES | CCCC1=C(C2=C(C=CC=N2)C=C1)O |
Chemical Identifiers
58327-60-9 | |
187.242 | |
OKPSVYHYNXVYED-UHFFFAOYSA-N | |
11604513 | |
CCCC1=C(C2=C(C=CC=N2)C=C1)O |
C12H13NO | |
MFCD00059755 | |
7-Propyloxine, 7-Propyl-8-quinolinol | |
7-propylquinolin-8-ol |
Specifications
149°C | |
0°C | |
58327-60-9 | |
MFCD00059755 | |
OKPSVYHYNXVYED-UHFFFAOYSA-N | |
7-propylquinolin-8-ol | |
11604513 | |
Clear Liquid at 20°C | |
8-Hydroxy-7-propylquinoline |
Undesignated | |
1g | |
C12H13NO | |
7-Propyloxine, 7-Propyl-8-quinolinol | |
CCCC1=C(C2=C(C=CC=N2)C=C1)O | |
187.242 | |
187.24 | |
≥98.0% (GC,T) |
Safety and Handling
TSCA : No