Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Hydroxy-7-methoxycoumarin, 97%

$369.16 - $369.16

Chemical Identifiers

Chemical Name or Material 8-Hydroxy-7-methoxycoumarin
CAS 19492-03-6
Molecular Formula C10H8O4
Formula Weight 192.17
MDL Number MFCD00075652
Beilstein 168393
Synonym Daphnetin 7-methyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL13841ME

  SDS
alfa aesar™
L13841ME
500mg Each for $369.16
Description & Specifications

Specifications

Formula Weight 192.17
Chemical Name or Material 8-Hydroxy-7-methoxycoumarin
MDL Number MFCD00075652
Health Hazard 1 Warning
CAS 19492-03-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Beilstein 168393
Melting Point 175° to 177°C
Quantity 500mg
Odor Odorless
Molecular Formula C10H8O4
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Synonym Daphnetin 7-methyl ether
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.