missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Hydroxy-7-methoxycoumarin, 97%

Manufacturer:  Alfa Aesar™ L13841ME

 View more versions of this product

Catalog No. AAL13841ME



Specifications

Specifications

8-Hydroxy-7-methoxycoumarin
175°C to 177°C
19492-03-6
MFCD00075652
8-hydroxy-7-methoxycoumarin, daphnetin-7-methylether, 8-hydroxy-7-methoxy-2h-chromen-2-one, 8-hydroxy-7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 8-hydroxy-7-methoxy, 2h-1-benzopyran-2-one,8-hydroxy-7-methoxy, acmc-20alwe, daphnetin 7-methylether, daphnetin 7-methyl ether, daphnetin-7-methyl ether
COC1=C(C2=C(C=C1)C=CC(=O)O2)O
192.17
192.17
Odorless
500mg
C10H8O4
168393
KIGCGZUAVODHMD-UHFFFAOYSA-N
8-hydroxy-7-methoxychromen-2-one
146487
97%

Chemical Identifiers

19492-03-6
192.17
KIGCGZUAVODHMD-UHFFFAOYSA-N
146487
COC1=C(C2=C(C=C1)C=CC(=O)O2)O
C10H8O4
MFCD00075652
8-hydroxy-7-methoxycoumarin, daphnetin-7-methylether, 8-hydroxy-7-methoxy-2h-chromen-2-one, 8-hydroxy-7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 8-hydroxy-7-methoxy, 2h-1-benzopyran-2-one,8-hydroxy-7-methoxy, acmc-20alwe, daphnetin 7-methylether, daphnetin 7-methyl ether, daphnetin-7-methyl ether
8-hydroxy-7-methoxychromen-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents