Please login to your online account to display your discounted pricing

8-Hydroxy-5-nitroquinoline, 96%, Acros Organics


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  161201000

 View more versions of this product

Catalog No. AC161201000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥95%
Formula Weight 190.16
Color Ochre-yellow to brownish
Quantity 100g
Merck Index 15, 6742
EINECS Number 223-662-4
RTECS Number VC8225000
Melting Point 177° to 183°C
CAS Max % 100.0
CAS Min % 95.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 8-Hydroxy-5-nitroquinoline
Name Note 96%
Assay Percent Range 96%
Molecular Formula C9H6N2O3
MDL Number MFCD00006791
CAS 4008-48-4
Synonym 5-Nitro-8-quinolinol

Specifications

Chemical Name or Material 8-Hydroxy-5-nitroquinoline
Name Note 96%
CAS 4008-48-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C9H6N2O3
Formula Weight 190.16
MDL Number MFCD00006791
Synonym 5-Nitro-8-quinolinol
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents