missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Hydroxy-5-nitroquinoline, 96%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 161200250

 View more versions of this product

Catalog No. AC161200250


 

Specifications Description & Specifications

Specifications

≥95%
Crystalline Powder
25g
15, 6742
100.0
Authentic
96%
C9H6N2O3
96%
190.16
Ochre-yellow to brownish
177° to 183°C
95.0
5-Nitro-8-quinolinol
8-Hydroxy-5-nitroquinoline
4008-48-4
MFCD00006791

Chemical Identifiers

4008-48-4
100.0
C9H6N2O3
MFCD00006791
8-Hydroxy-5-nitroquinoline, 96%
95.0
96%
190.16
5-Nitro-8-quinolinol
≥95%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 223-662-4

RTECSNumber : VC8225000