Please login to your online account to display your discounted pricing

8-Hydroxy-5-nitroquinoline, 96%, Acros Organics

$92.99 - $92.99

Chemical Identifiers

Chemical Name or Material 8-Hydroxy-5-nitroquinoline
Name Note 96%
CAS 4008-48-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C9H6N2O3
Formula Weight 190.16
MDL Number MFCD00006791
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC161200250

  SDS
thermo fisher scientific
161200250
Each for $92.99

AC161201000

  SDS
thermo fisher scientific
161201000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Formula Weight 190.16
Physical Form Crystalline Powder
Color Ochre-yellow to brownish
Quantity 25g
EINECS Number 223-662-4
RTECS Number VC8225000
Melting Point 177° to 183°C
Merck Index 15, 6742
CAS Min % 95.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Synonym 5-Nitro-8-quinolinol
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 8-Hydroxy-5-nitroquinoline
Assay Percent Range 96%
CAS 4008-48-4
Molecular Formula C9H6N2O3
MDL Number MFCD00006791
Name Note 96%
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.