Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Hydroxy-5-nitroquinoline, 96%

$42.80 - $248.62

Chemical Identifiers

Chemical Name or Material 8-Hydroxy-5-nitroquinoline
CAS 4008-48-4
Molecular Formula C9H6N2O3
Formula Weight 190.16
MDL Number MFCD00006791
Merck Index 14,6655
Synonym 5-Nitro-8-quinolinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2446206

  SDS
alfa aesar™
B2446206
5g Each for $42.80

AAB2446214

  SDS
alfa aesar™
B2446214
25g Each for $100.96

AAB2446222

  SDS
alfa aesar™
B2446222
100g Each for $248.62
Description & Specifications

Specifications

Formula Weight 190.16
Chemical Name or Material 8-Hydroxy-5-nitroquinoline
Assay 96%
Molecular Formula C9H6N2O3
MDL Number MFCD00006791
CAS 4008-48-4
Merck Index 14,6655
Synonym 5-Nitro-8-quinolinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VC8225000
Odor Odorless
Melting Point 177° to 182°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.