Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Hexadecanol, 97%

$118.99 - $1,633.18

Chemical Identifiers

Chemical Name or Material 8-Hexadecanol
CAS 19781-83-0
Molecular Formula CH3(CH2)7CH(OH)(CH2)6CH3
Formula Weight 242.45
MDL Number MFCD00046733
Synonym n-Heptyl n-octyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2073406

  SDS
alfa aesar™
B2073406
5g Each for $118.99

AAB2073422

  SDS
alfa aesar™
B2073422
100g Each for $1,633.18

AAB2073414

  SDS
alfa aesar™
B2073414
25g Each for $474.09
Description & Specifications

Specifications

Formula Weight 242.45
Chemical Name or Material 8-Hexadecanol
Assay 97%
Molecular Formula CH3(CH2)7CH(OH)(CH2)6CH3
MDL Number MFCD00046733
CAS 19781-83-0
Synonym n-Heptyl n-octyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g

Applications
It is employed in the synthesis and absolute configuration of (+)-8-hydroxyhexadecanoic acid, an endogenous inhibitor for spore germination in Lygodium japonicum.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.