Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Ethoxyquinoline-5-sulfonic acid sodium salt hydrate, 97%

$22.61 - $355.59

Chemical Identifiers

Chemical Name or Material 8-Ethoxyquinoline-5-sulfonic acid sodium salt hydrate
CAS 80789-76-0
Molecular Formula C11H10NNaO4S·xH2O
Formula Weight 275.26 (Anhydrous)
MDL Number MFCD00149478
Synonym Actinoquinol; Sodium 8-ethoxyquinoline-5-sulfonate hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1666809

 
alfa aesar™
A1666809
10g Each for $22.61

AAA1666818

 
alfa aesar™
A1666818
50g Each for $77.15

AAA1666830

 
alfa aesar™
A1666830
250g Each for $355.59
Description & Specifications

Specifications

Formula Weight 275.26 (Anhydrous)
Quantity 10g
Chemical Name or Material 8-Ethoxyquinoline-5-sulfonic acid sodium salt hydrate
Molecular Formula C11H10NNaO4S·xH2O
CAS 80789-76-0
MDL Number MFCD00149478
Melting Point >280°C
Assay 97%
Synonym Actinoquinol; Sodium 8-ethoxyquinoline-5-sulfonate hydrate