missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-(Diethylamino)octyl 3,4,5-trimethoxybenzoate hydrochloride, 97%

Inhibits intracellular calcium mobilization

$172.72 - $794.19

Chemical Identifiers

CAS 53464-72-5
Molecular Formula C22H38ClNO5
Molecular Weight (g/mol) 431.998
MDL Number MFCD00012502
InChI Key KFJZVXKPPQIYCG-UHFFFAOYSA-N
Synonym 8-diethylamino octyl 3,4,5-trimethoxybenzoate hydrochloride, tmb-8 hydrochloride, tmb-8, hydrochloride, 8-diethylamino octyl-3,4,5-trimethoxybenzoate, 8-n,n-diethylamino-octyl-3,4,5-trimethoxybenzoate, hcl, 8-diethylamino octyl-3,4,5-trimethoxybenzoate hydrochloride, 3,4,5-trimethoxybenzoic acid 8-diethylamino octyl ester, hydrochloride, tmb 8 hydrochloride, acmc-1b069, regid_for_cid_6917843
PubChem CID 6917843
IUPAC Name 8-(diethylamino)octyl 3,4,5-trimethoxybenzoate;hydrochloride
SMILES CCN(CC)CCCCCCCCOC(=O)C1=CC(=C(C(=C1)OC)OC)OC.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62528MF View Documents Alfa Aesar™
J62528MF
50mg Each for $172.72
 
AAJ62528MD View Documents Alfa Aesar™
J62528MD
250mg Each for $794.19
 
Specifications

Chemical Identifiers

53464-72-5
431.998
KFJZVXKPPQIYCG-UHFFFAOYSA-N
6917843
CCN(CC)CCCCCCCCOC(=O)C1=CC(=C(C(=C1)OC)OC)OC.Cl
C22H38ClNO5
MFCD00012502
8-diethylamino octyl 3,4,5-trimethoxybenzoate hydrochloride, tmb-8 hydrochloride, tmb-8, hydrochloride, 8-diethylamino octyl-3,4,5-trimethoxybenzoate, 8-n,n-diethylamino-octyl-3,4,5-trimethoxybenzoate, hcl, 8-diethylamino octyl-3,4,5-trimethoxybenzoate hydrochloride, 3,4,5-trimethoxybenzoic acid 8-diethylamino octyl ester, hydrochloride, tmb 8 hydrochloride, acmc-1b069, regid_for_cid_6917843
8-(diethylamino)octyl 3,4,5-trimethoxybenzoate;hydrochloride

Specifications

White
50mg
C22H38ClNO5
5466611
KFJZVXKPPQIYCG-UHFFFAOYSA-N
8-(diethylamino)octyl 3,4,5-trimethoxybenzoate;hydrochloride
6917843
Powder
8-(Diethylamino)octyl 3,4,5-trimethoxybenzoate hydrochloride
92°C to 97°C
53464-72-5
MFCD00012502
8-diethylamino octyl 3,4,5-trimethoxybenzoate hydrochloride, tmb-8 hydrochloride, tmb-8, hydrochloride, 8-diethylamino octyl-3,4,5-trimethoxybenzoate, 8-n,n-diethylamino-octyl-3,4,5-trimethoxybenzoate, hcl, 8-diethylamino octyl-3,4,5-trimethoxybenzoate hydrochloride, 3,4,5-trimethoxybenzoic acid 8-diethylamino octyl ester, hydrochloride, tmb 8 hydrochloride, acmc-1b069, regid_for_cid_6917843
CCN(CC)CCCCCCCCOC(=O)C1=CC(=C(C(=C1)OC)OC)OC.Cl
431.998
432
97%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C