missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Cyclopentyl-1,3-dimethylxanthine

A selective A1 adenosine receptor antagonist

$240.57 - $508.91

Chemical Identifiers

CAS 35873-49-5
Molecular Formula C12H16N4O2
Molecular Weight (g/mol) 248.286
MDL Number MFCD00055116
InChI Key SCVHFRLUNIOSGI-UHFFFAOYSA-N
Synonym 8-cyclopentyltheophylline, 8-cyclopentyl-1,3-dimethylxanthine, 8-cyclopentyl theophylline, 1,3-dimethyl-8-cyclopentylxanthine, xanthine, 8-cyclopentyl-1,3-dimethyl, theophylline, 8-cyclopentyl, 8-cyclopentyl-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6-dione, 8-cyclopentyl-3,9-dihydro-1,3-dimethyl, spectrum_000351, lopac-c-102
PubChem CID 1917
IUPAC Name 8-cyclopentyl-1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)NC(=N2)C3CCCC3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61565MC View Documents Alfa Aesar™
J61565MC
100mg Each for $240.57
 
AAJ61565MD View Documents Alfa Aesar™
J61565MD
250mg Each for $508.91
 
Specifications

Chemical Identifiers

35873-49-5
248.286
SCVHFRLUNIOSGI-UHFFFAOYSA-N
1917
CN1C2=C(C(=O)N(C1=O)C)NC(=N2)C3CCCC3
C12H16N4O2
MFCD00055116
8-cyclopentyltheophylline, 8-cyclopentyl-1,3-dimethylxanthine, 8-cyclopentyl theophylline, 1,3-dimethyl-8-cyclopentylxanthine, xanthine, 8-cyclopentyl-1,3-dimethyl, theophylline, 8-cyclopentyl, 8-cyclopentyl-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6-dione, 8-cyclopentyl-3,9-dihydro-1,3-dimethyl, spectrum_000351, lopac-c-102
8-cyclopentyl-1,3-dimethyl-7H-purine-2,6-dione

Specifications

White
100mg
C12H16N4O2
8-cyclopentyltheophylline, 8-cyclopentyl-1,3-dimethylxanthine, 8-cyclopentyl theophylline, 1,3-dimethyl-8-cyclopentylxanthine, xanthine, 8-cyclopentyl-1,3-dimethyl, theophylline, 8-cyclopentyl, 8-cyclopentyl-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6-dione, 8-cyclopentyl-3,9-dihydro-1,3-dimethyl, spectrum_000351, lopac-c-102
SCVHFRLUNIOSGI-UHFFFAOYSA-N
8-cyclopentyl-1,3-dimethyl-7H-purine-2,6-dione
1917
Powder
250°C to 252°C
35873-49-5
MFCD00055116
Soluble in 0.1M NaOH; Slightly soluble in water; Also soluble in DMSO at 16mg/ml at 60°C
CN1C2=C(C(=O)N(C1=O)C)NC(=N2)C3CCCC3
248.286
248.28
8-Cyclopentyl-1,3-dimethylxanthine
Safety and Handling

Safety and Handling

RTECSNumber : XH5093600

Recommended Storage : 2° to 8°C