missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Cyclopentyl-1,3-dimethylxanthine

A selective A1 adenosine receptor antagonist

Manufacturer:  Alfa Aesar™ J61565MD

 View more versions of this product

Catalog No. AAJ61565MD



Specifications

Specifications

White
250mg
C12H16N4O2
1,3-dimethyl-8-cyclopentylxanthine, 1h-purine-2,6-dione, 8-cyclopentyl-3,9-dihydro-1,3-dimethyl, 8-cyclopentyl theophylline, 8-cyclopentyl-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 8-cyclopentyl-1,3-dimethylxanthine, 8-cyclopentyltheophylline, lopac-c-102, spectrum_000351, theophylline, 8-cyclopentyl, xanthine, 8-cyclopentyl-1,3-dimethyl
SCVHFRLUNIOSGI-UHFFFAOYSA-N
8-cyclopentyl-1,3-dimethyl-7H-purine-2,6-dione
1917
Powder
250°C to 252°C
35873-49-5
MFCD00055116
Soluble in 0.1M NaOH; Slightly soluble in water; Also soluble in DMSO at 16mg/ml at 60°C
CN1C2=C(C(=O)N(C1=O)C)NC(=N2)C3CCCC3
248.286
248.28

Chemical Identifiers

35873-49-5
248.286
SCVHFRLUNIOSGI-UHFFFAOYSA-N
1917
CN1C2=C(C(=O)N(C1=O)C)NC(=N2)C3CCCC3
C12H16N4O2
MFCD00055116
1,3-dimethyl-8-cyclopentylxanthine, 1h-purine-2,6-dione, 8-cyclopentyl-3,9-dihydro-1,3-dimethyl, 8-cyclopentyl theophylline, 8-cyclopentyl-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 8-cyclopentyl-1,3-dimethylxanthine, 8-cyclopentyltheophylline, lopac-c-102, spectrum_000351, theophylline, 8-cyclopentyl, xanthine, 8-cyclopentyl-1,3-dimethyl
8-cyclopentyl-1,3-dimethyl-7H-purine-2,6-dione
Safety and Handling

Safety and Handling

RTECSNumber : XH5093600

Recommended Storage : 2° to 8°C

Documents