Please login to your online account to display your discounted pricing

8 Component Mix in DMSO, SPEX CertiPrep™


Quantity 1mL
Packaging Glass Ampoule

Manufacturer: spex certiprep llc  USPRSC2B

 View more versions of this product

Catalog No. USPRSC2B

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1mL
Density 1.1g/cm3
CAS 127-19-5
Name Note USP 467 Standard
Chemical Name or Material 8 Component Mix in DMSO
Physical Form Fluid
CAS Various Additional
Purity Grade USP
Tetramethylene sulfone 800μg/mL
Solvent or Matrix DMSO
Methods USP 467: Residual Solvents/ Organic Volatile Impurities
Packaging Glass Ampoule
CAS 75-12-7
Analyte or Component Names N,N-Dimethylacetamide; N,N-Dimethylformamide; Ethylene glycol; 1-Methyl-2-pyrrolidinone; Formamide; more
N,N_Dimethylacetamide 5450μg/mL
Formamide 1100μg/mL
2_Methoxyethanol 250μg/mL
Ignition Point 270°C
CAS 107-21-1
2_Ethoxyethanol 800μg/mL
Vapor Pressure 2.5 hPA (20°C)
CAS 68-12-2
1_Methyl_2_pyrrolidinone 2650μg/mL
Ethylene glycol 3100μg/mL
Explosion Limit 1.8% (volume); 63% (volume)
CAS 872-50-4
Recommended Storage Freezer

Specifications

Chemical Name or Material 8 Component Mix in DMSO
Name Note USP 467 Standard
CAS 127-19-5
CAS 68-12-2
CAS 107-21-1
CAS 872-50-4
CAS 75-12-7
CAS Various Additional
Purity Grade USP
Accessories

Freezer