Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Chlorotheophylline, 99%

$45.81 - $589.15

Chemical Identifiers

Chemical Name or Material 8-Chlorotheophylline
CAS 85-18-7
Molecular Formula C7H7ClN4O2
Formula Weight 214.61
MDL Number MFCD00005581
Beilstein 203068
Synonym 8-Chloro-1,3-dimethyl-2,6-purinedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA124080B

  SDS
alfa aesar™
A124080B
1000g Each for $589.15

AAA1240818

  SDS
alfa aesar™
A1240818
50g Each for $45.81

AAA1240830

  SDS
alfa aesar™
A1240830
250g Each for $173.28
Description & Specifications

Specifications

Formula Weight 214.61
Molecular Formula C7H7ClN4O2
MDL Number MFCD00005581
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Melting Point ≈290°C (dec.)
Assay 99%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XH5063000
Quantity 1000g
CAS 85-18-7
Synonym 8-Chloro-1,3-dimethyl-2,6-purinedione
Chemical Name or Material 8-Chlorotheophylline
Beilstein 203068

Applications
8-Chlorotheophylline is a xanthine alkaloid derivative. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in sodium hydroxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.