Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Chloronaphthalene-1-sulfonic acid dihydrate, 97%

Chemical Identifiers

Chemical Name or Material 8-Chloronaphthalene-1-sulfonic acid dihydrate
CAS 1171630-97-9
Molecular Formula C10H7ClO3S·2H2O
Formula Weight 278.71 (242.69 Anhydrous)
MDL Number MFCD00052379
UN Number UN2585
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2094809

  SDS
alfa aesar™
B2094809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2094888

  SDS
alfa aesar™
B2094888
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 278.71 (242.69 Anhydrous)
TSCA TSCA
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material 8-Chloronaphthalene-1-sulfonic acid dihydrate
Assay 97%
Molecular Formula C10H7ClO3S·2H2O
MDL Number MFCD00052379
CAS 1171630-97-9
UN Number UN2585
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
Melting Point 145° to 147°C
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.