missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-chloronaphthalene-1-sulfonic acid, 97%, Maybridge

Manufacturer:  Fisher Scientific Company BTB06262EA

 View more versions of this product

Catalog No. BTB06262EA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

8-chloronaphthalene-1-sulfonic acid
96.5
Amber glass bottle
97%
C10H7ClO3S
MFCD00052379
145-74-4
100.0
10g
97%
242.68

Chemical Identifiers

145-74-4
100.0
C10H7ClO3S
MFCD00052379
97%
96.5
97%
242.68
8-chloronaphthalene-1-sulfonic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger