missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one, TCI America™

Manufacturer:  TCI America C31815G

 View more versions of this product

Catalog No. C31815G



Specifications

Specifications

8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one
105°C
31251-41-9
MFCD00800222
WMQNOYVVLMIZDV-UHFFFAOYSA-N
8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-one
9816260
Crystal-Powder at 20°C
White-Yellow
5g
C14H10ClNO
8-Chloro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11(6H)-one
C1CC2=C(C=CC(=C2)Cl)C(=O)C3=C1C=CC=N3
243.69
243.69
≥98.0% (GC,T)

Chemical Identifiers

31251-41-9
243.69
WMQNOYVVLMIZDV-UHFFFAOYSA-N
9816260
C1CC2=C(C=CC(=C2)Cl)C(=O)C3=C1C=CC=N3
C14H10ClNO
MFCD00800222
8-Chloro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11(6H)-one
8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents