Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline, 97%

$70.19 - $70.19

Chemical Identifiers

Chemical Name or Material 8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline
CAS 18706-22-4
Molecular Formula C10H5ClF3NO
Formula Weight 247.61
MDL Number MFCD00276589
Synonym 8-Chloro-2-(trifluoromethyl)-4-quinolinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1989203

 
alfa aesar™
A1989203
1g Each for $70.19
Description & Specifications

Specifications

Formula Weight 247.61
Chemical Name or Material 8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline
Assay 97%
Molecular Formula C10H5ClF3NO
MDL Number MFCD00276589
CAS 18706-22-4
Synonym 8-Chloro-2-(trifluoromethyl)-4-quinolinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 138° to 140°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.