missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Chloro-1,2,4-triazolo[4,3-a]pyrazine, 97%

Manufacturer:  Alfa Aesar™ H63343MD

 View more versions of this product

Catalog No. AAH63343MD



Specifications

Specifications

8-Chloro-1,2,4-triazolo[4,3-a]pyrazine
215°C to 220°C
68774-77-6
MFCD08272086
VKZJRJVVFDDJKP-UHFFFAOYSA-N
8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
12582612
Solid
Brown
250mg
C5H3ClN4
8-chloro-1,2,4 triazolo 4,3-a pyrazine, 8-chloro 1,2,4 triazolo 4,3-a pyrazine, 8-chlorotriazolo 4,3-a pyrazine, 1,2,4-triazolo 4,3-a pyrazine, 8-chloro, 8-chloro-1,2,4triazolo 4,3-a pyrazine, 1,2,4-triazolo 4,3-a pyrazine,8-chloro, 8-chloro-1,2,4-triazolo 4,3-a pyrazine
C1=CN2C=NN=C2C(=N1)Cl
154.557
154.56
97%

Chemical Identifiers

68774-77-6
154.557
VKZJRJVVFDDJKP-UHFFFAOYSA-N
12582612
C1=CN2C=NN=C2C(=N1)Cl
C5H3ClN4
MFCD08272086
8-chloro-1,2,4 triazolo 4,3-a pyrazine, 8-chloro 1,2,4 triazolo 4,3-a pyrazine, 8-chlorotriazolo 4,3-a pyrazine, 1,2,4-triazolo 4,3-a pyrazine, 8-chloro, 8-chloro-1,2,4triazolo 4,3-a pyrazine, 1,2,4-triazolo 4,3-a pyrazine,8-chloro, 8-chloro-1,2,4-triazolo 4,3-a pyrazine
8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents