Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Carboxynaphthalene-1-carboxamide, tech. 85%

$101.57 - $101.57

Chemical Identifiers

Chemical Name or Material 8-Carboxynaphthalene-1-carboxamide
CAS 5811-88-1
Purity Grade Technical Grade
Molecular Formula C12H9NO3
Formula Weight 215.21
MDL Number MFCD00052020
Beilstein 2213597
Synonym 8-Carbamoylnaphthalene-1-carboxylic acid; 1,8-Naphthalic acid monoamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1262806

  SDS
alfa aesar™
L1262806
5g Each for $101.57
Description & Specifications

Specifications

Formula Weight 215.21
Chemical Name or Material 8-Carboxynaphthalene-1-carboxamide
MDL Number MFCD00052020
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Purity Grade Technical Grade
Molecular Formula C12H9NO3
CAS 5811-88-1
Beilstein 2213597
Synonym 8-Carbamoylnaphthalene-1-carboxylic acid; 1,8-Naphthalic acid monoamide
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Assay 85%
Melting Point 265° to 270°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.