missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Carboxynaphthalene-1-carboxamide, tech. 85%

Manufacturer:  Alfa Aesar™ L1262814

 View more versions of this product

Catalog No. AAL1262814



Specifications

Specifications

8-Carboxynaphthalene-1-carboxamide
25g
C12H9NO3
2213597
GRVIMBYDMARKEZ-UHFFFAOYSA-N
8-carbamoylnaphthalene-1-carboxylic acid
2777172
85%
265°C to 270°C
5811-88-1
MFCD00052020
8-aminocarbonyl-1-naphthoic acid, 8-carboxynaphthalene-1-carboxamide, 8-carbamoyl-1-naphthoic acid, 8-carbamoylnaphthalenecarboxylic acid, maybridge1_003186, acmc-1am92, 1,8-naphthalenedicarboxylic imide, 8-carboxynaphyhalene-1-carboxamide, 8-carbamoyl-1-naphthalenecarboxylic acid, 8-aminocarbonylnaphthalene-1-carboxylic acid
C1=CC2=C(C(=C1)C(=O)N)C(=CC=C2)C(=O)O
215.208
215.21
Technical

Chemical Identifiers

5811-88-1
215.208
GRVIMBYDMARKEZ-UHFFFAOYSA-N
2777172
C1=CC2=C(C(=C1)C(=O)N)C(=CC=C2)C(=O)O
C12H9NO3
MFCD00052020
8-aminocarbonyl-1-naphthoic acid, 8-carboxynaphthalene-1-carboxamide, 8-carbamoyl-1-naphthoic acid, 8-carbamoylnaphthalenecarboxylic acid, maybridge1_003186, acmc-1am92, 1,8-naphthalenedicarboxylic imide, 8-carboxynaphyhalene-1-carboxamide, 8-carbamoyl-1-naphthalenecarboxylic acid, 8-aminocarbonylnaphthalene-1-carboxylic acid
8-carbamoylnaphthalene-1-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents