missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Bromoquinoline, 97%, Maybridge

Chemical Identifiers

CAS 16567-18-3
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD00191859
InChI Key PIWNKSHCLTZKSZ-UHFFFAOYSA-N
Synonym 8-bromo-quinoline
PubChem CID 140109
IUPAC Name 8-bromoquinoline
SMILES C1=CC2=C(C(=C1)Br)N=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC04310DA SDS View Documents Fisher Scientific Company
CC04310DA
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC04310EA SDS View Documents Fisher Scientific Company
CC04310EA
10g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC04310EB SDS View Documents Fisher Scientific Company
CC04310EB
25g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

16567-18-3
208.058
PIWNKSHCLTZKSZ-UHFFFAOYSA-N
140109
C1=CC2=C(C(=C1)Br)N=CC=C2
C9H6BrN
MFCD00191859
8-bromo-quinoline
8-bromoquinoline

Specifications

1.5940g/mL
1g
96.5
97%
MFCD00191859
PIWNKSHCLTZKSZ-UHFFFAOYSA-N
8-bromoquinoline
140109
97%
Amber glass bottle
16567-18-3
100.0
C9H6BrN
8-bromo-quinoline
C1=CC2=C(C(=C1)Br)N=CC=C2
208.058
208.06
8-Bromoquinoline
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.