missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Bromoquinoline, 97%, Maybridge

Manufacturer:  Thermo Scientific CC04310EB

 View more versions of this product

Catalog No. CC04310EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

8-Bromoquinoline
Amber glass bottle
16567-18-3
100.0
C9H6BrN
quinoline, 8-bromo, 8-bromo-quinoline, 8-bromoquinoline, pubchem5816, 8-bromquinoline, 8-bromoquinoline, acmc-209dt3, ksc181q5t, buttpark 33\04-57
C1=CC2=C(C(=C1)Br)N=CC=C2
208.058
208.06
1.5940g/mL
25g
96.5
97%
MFCD00191859
PIWNKSHCLTZKSZ-UHFFFAOYSA-N
8-bromoquinoline
140109
97%

Chemical Identifiers

16567-18-3
208.058
PIWNKSHCLTZKSZ-UHFFFAOYSA-N
140109
C1=CC2=C(C(=C1)Br)N=CC=C2
C9H6BrN
MFCD00191859
quinoline, 8-bromo, 8-bromo-quinoline, 8-bromoquinoline, pubchem5816, 8-bromquinoline, 8-bromoquinoline, acmc-209dt3, ksc181q5t, buttpark 33\04-57
8-bromoquinoline
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Documents