missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Bromoimidazo[1,2-a]pyridine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446610010

 View more versions of this product

Catalog No. AC446610010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

8-Bromoimidazo[1, 2-a]pyridine
72°C to 75°C
850349-02-9
C7H5BrN2
GCCVNNZARPFXES-UHFFFAOYSA-N
8-bromoimidazo[1,2-a]pyridine
17749933
95%
Glass Bottle
1g
94% min. (GC)
8-bromo-imidazo 1,2-a pyridine, 8-bromoh-imidazo 1,2-a pyridine, 8-bromoimidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 8-bromo, imidazo 1,2-a pyridine,8-bromo
C1=CN2C=CN=C2C(=C1)Br
197.03
197.03

Chemical Identifiers

850349-02-9
197.03
8-bromo-imidazo 1,2-a pyridine, 8-bromoh-imidazo 1,2-a pyridine, 8-bromoimidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 8-bromo, imidazo 1,2-a pyridine,8-bromo
8-bromoimidazo[1,2-a]pyridine
C7H5BrN2
GCCVNNZARPFXES-UHFFFAOYSA-N
17749933
C1=CN2C=CN=C2C(=C1)Br
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and ke

GHS Signal Word: Warning

SDS
Documents