missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Bromoimidazo[1,2-a]pyridine, 95%, ACROS Organics™
Chemical Identifiers
CAS | 850349-02-9 |
---|---|
Molecular Formula | C7H5BrN2 |
Molecular Weight (g/mol) | 197.03 |
InChI Key | GCCVNNZARPFXES-UHFFFAOYSA-N |
Synonym | 8-bromo-imidazo 1,2-a pyridine, 8-bromoh-imidazo 1,2-a pyridine, 8-bromoimidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 8-bromo, imidazo 1,2-a pyridine,8-bromo |
PubChem CID | 17749933 |
IUPAC Name | 8-bromoimidazo[1,2-a]pyridine |
SMILES | C1=CN2C=CN=C2C(=C1)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC446610010
|
Thermo Fisher Scientific
AC446610010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC446610050
|
Thermo Fisher Scientific
AC446610050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
850349-02-9 | |
197.03 | |
8-bromo-imidazo 1,2-a pyridine, 8-bromoh-imidazo 1,2-a pyridine, 8-bromoimidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 8-bromo, imidazo 1,2-a pyridine,8-bromo | |
8-bromoimidazo[1,2-a]pyridine |
C7H5BrN2 | |
GCCVNNZARPFXES-UHFFFAOYSA-N | |
17749933 | |
C1=CN2C=CN=C2C(=C1)Br |
Specifications
Glass Bottle | |
1g | |
94% min. (GC) | |
8-bromo-imidazo 1,2-a pyridine, 8-bromoh-imidazo 1,2-a pyridine, 8-bromoimidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 8-bromo, imidazo 1,2-a pyridine,8-bromo | |
C1=CN2C=CN=C2C(=C1)Br | |
197.03 | |
197.03 | |
8-Bromoimidazo[1, 2-a]pyridine |
72°C to 75°C | |
850349-02-9 | |
C7H5BrN2 | |
GCCVNNZARPFXES-UHFFFAOYSA-N | |
8-bromoimidazo[1,2-a]pyridine | |
17749933 | |
95% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and ke
GHS Signal Word: Warning